Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TransMoDE 3 Biotin

Catalog No. 8829100U
Change view
Click to view available options
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
88-291-00U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-291-00U Supplier Tocris Bioscience™ Supplier No. 8829/100U
Only null left
Add to Cart
Add to Cart

Biotinylated angiopep-2, induces internalization and degradation of streptavidin in brain cell lines

TransMoDE 3 Biotin is a biotinylated peptide capable of inducing both the degradation of target protein via lysosomal proteolysis and their bidirectional transcytosis of streptavidin across the blood-brain barrier. TransMoDE 3 Biotin is derived from the peptide Angiopep-2 (Cat. No. 8828).

Specifications

Purity 95%
Content And Storage Store at -20°C
Molecular Weight (g/mol) Observed: 2611.88
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.