Learn More
LGC GENOMICS LLC (Lucigen) TransforMax EC100D pir-116 Electrocompetent E. coli, 10 RxnsPerform plasmid rescue for transposition mutants created with the EZ-Tn5 Transposome Kits.
Catalog No. NC9801351
Encompass
These cells are used for rescue cloning of DNA transposed using the EZ-Tn5™ Insertion kit or EZ-Tn5™ TNP Transposome™. The EC100D pir-116 cells enable propagation of up to 250 rescue plasmid copies per cell.
This item is not returnable. View return policy
Catalog No. NC9801351 Supplier LGC GENOMICS LLC Supplier No. EC6P095H
These cells are used for rescue cloning of DNA transposed using the EZ-Tn5™ Insertion kit or EZ-Tn5™ TNP Transposome™. The EC100D pir-116 cells enable propagation of up to 250 rescue plasmid copies per cell.