Learn More
LGC GENOMICS LLC (Lucigen) TransforMax Ec100D pir+ Electrocompetent E. coli, 10 RxnsPerform plasmid rescue for transposition mutants created with the EZ-Tn5 Transposome Kits.
Catalog No. NC1342337
Encompass
These cells are used for rescue cloning of DNA transposed using the EZ-Tn5™ Insertion kit or EZ-Tn5™ TNP Transposome™. The EC100D pir+ cells enable propagation of about 15 rescue plasmid copies per cell.
This item is not returnable. View return policy
Catalog No. NC1342337 Supplier LGC GENOMICS LLC Supplier No. ECP09500
These cells are used for rescue cloning of DNA transposed using the EZ-Tn5™ Insertion kit or EZ-Tn5™ TNP Transposome™. The EC100D pir+ cells enable propagation of about 15 rescue plasmid copies per cell.