Learn More
Agilent Technologies TRANSPACK PACKAGING EXTRACT
Supplier:  Agilent Technologies 200223
Encompass
Transpack Packaging Extract, Ideal for recovery of all lambda shuttle vectors, No artifacts from background B-gal, Highest efficiency packaging extract available, Deficient in McrA, McrBC, Mrr and McrF restriction systems
Catalog No. 50-125-041