Learn More
Agilent Technologies TRANSPACK PACKAGING EXTRACT
Catalog No. 50125040
Encompass
Transpack Packaging Extract 100 vials Ideal for recovery of all lambda shuttle vectors Lack ofbeta-gal activity No artifacts affecting mutation data Highest recovery per extract reaction Deficient in McrA McrBC Mrr and McrF
Catalog No. 50-125-040 Supplier Agilent Technologies Supplier No. 200221
Transpack Packaging Extract 100 vials Ideal for recovery of all lambda shuttle vectors Lack ofbeta-gal activity No artifacts affecting mutation data Highest recovery per extract reaction Deficient in McrA McrBC Mrr and McrF