Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erlab™ Transparent Back Panel For Captair 483 Fume Hood
GREENER_CHOICE

Catalog No. 08444028 Shop All Erlab Products
Change view
Click to view available options
Material:
Acrylic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
08-444-028 Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-444-028 Supplier Erlab™ Supplier No. 483EDU
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clear back panel made of synthetic glass offering 360° visibility for 483 fume hood models.

  • For use with Captair 483 Ductless Filtering Fume Hood
  • Includes warranty
  • Useful for teaching and student observation

Specifications

For Use With (Equipment) Captair Ductless Filtering Fume Hoods or CaptairFlow Laminar Flow Hoods
Weight (English) 28 lb.
Weight (Metric) 12.7 Kg
Includes Educational Back Panel and Thumb Screws
Product Type Back Panel
Voltage 0/1 V
Material Acrylic
For Use With (Application) Teaching Observation or Locating in Front of Window
Color Clear
Cut Style Rounded Edge
Warranty One Year
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.