Learn More
Ted Pella Inc Cryogenic Tape, 19mm x 15M, Roll, White Transparent
SDP
Catalog No. NC1874218
Encompass
Cryogenic tape for identification of already frozen vials, tubes, freezer boxes, plastic bags and other containers. Designed for long term bio preservation in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers. Not recommended for incubation in water baths and solvents. Overlap for best performance. 0.75" W x 50 ft. (19mm x 15M). Permanent and transparent.
Catalog No. NC1874218 Supplier Ted Pella Inc Supplier No. 223164
Cryogenic tape for identification of already frozen vials, tubes, freezer boxes, plastic bags and other containers. Designed for long term bio preservation in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers. Not recommended for incubation in water baths and solvents. Overlap for best performance. 0.75" W x 50 ft. (19mm x 15M). Permanent and transparent.