Learn More
GA International Transparent Dymo-Compatible Deep-Freeze Labels (Patent Pending) 0.5" x 1" / 12.7mm x 25.4mm Rectangle Transparent From -80°C to +70°C (-112°F to +158°F)
Supplier:  GA International EDFT040
Encompass_Preferred
Our DYMO-Compatible solvent-resistant deep-freeze labels are designed to identify containers destined for short or long-term storage in freezers and refrigerators. They are ideal for various surfaces and containers, including vials, microtubes, Falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. These freezer-grade labels are provided in a transparent format allowing the containers’ contents to remain visible at all times. Accordingly, they are recommended for containers with precious samples requiring monitoring, like tubes or microtubes containing blood and serum samples. As they also tolerate relatively high temperatures, up to +70°C (+158°F), these clear labels are also suitable for identifying cell culture flasks and plates stored in incubators (+37°C/+99°F). These direct thermal labels come with a permanent adhesive that will keep the label from falling off or flagging over time, perfect for samples stored for extended periods in
Catalog No. 50-005-70920