Learn More
JDM LAB SOLUTIONS LLC Crystal Transparent Lid, For DBC-06A Mini Dry Bath, 5.7 x 4.1 x 4.8 in, 1 ea
Catalog No. NC3933568
Encompass
This transparent lid is designed for use with the heating and cooling mini dry bath system. It provides visual access to samples during operation while helping to maintain consistent thermal conditions within the unit.
  • Provides a clear view of samples during incubation
  • Dimensions measure 5.7 x 4.1 x 4.8 in. (145 x 105 x 122.5 mm)
Catalog No. NC3933568 Supplier JDM LAB SOLUTIONS LLC Supplier No. DBCL03
This transparent lid is designed for use with the heating and cooling mini dry bath system. It provides visual access to samples during operation while helping to maintain consistent thermal conditions within the unit.
  • Provides a clear view of samples during incubation
  • Dimensions measure 5.7 x 4.1 x 4.8 in. (145 x 105 x 122.5 mm)