Learn More
Velp Scientific Transparent Lid for ECODryBlock

Description
- Transparent lid forms a warm air layer around blocks
- Provides full air coverage to reduce heat loss
- Minimizes condensation
- Reduces temperature variation across samples
- Supports stable and uniform conditions
- Made from heat-resistant material
- Allows visual monitoring without removing the lid
- Maintains consistent thermal conditions
Specifications
Specifications
| Color | Transparent |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Velp Scientifica ECODryBlock, Dry Block Heater |
| Material | Plastic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.