Learn More
CELLTREAT Scientific Products Transparent Microplate Sealing Film, Non-sterile
SDP
Catalog No. 5000144325
Encompass_Preferred
Transparent Sealing Film protects multiple well microplates from contamination and evaporation, including edge effectEasily removable and replaceable, ideal for incubation periods and short-term storage
Catalog No. 50-001-44325 Supplier CELLTREAT Scientific Products Supplier No. 229143
Transparent Sealing Film protects multiple well microplates from contamination and evaporation, including edge effectEasily removable and replaceable, ideal for incubation periods and short-term storage