Learn More
GA International Transparent Permanent Cryogenic & Autoclave-Resistant Laser Labels 0.394" x 0.315" / 10mm x 8mm Rectangle Transparent From -196°C to +150°C (-321°F to +302°F)
Catalog No. 5000571726
Encompass_Preferred
Cryogenic thermoplastic transparent laser labels for long-term storage in liquid nitrogen and deep-freeze harsh environments for biomedical research and clinical laboratories. The labels are also resistant to steam autoclave sterilization and boiling water. These transparent cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.
Catalog No. 50-005-71726 Supplier GA International Supplier No. DFLT121
Cryogenic thermoplastic transparent laser labels for long-term storage in liquid nitrogen and deep-freeze harsh environments for biomedical research and clinical laboratories. The labels are also resistant to steam autoclave sterilization and boiling water. These transparent cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.