Learn More
GA International Transparent Removable Cryogenic Labels for Laser Printers 1.42" x 0.55" / 36.1mm x 14mm Rectangle Transparent From -196°C to +120°C (-321°F to +248°F)
Catalog No. 5000571867
Encompass_Preferred
Transparent cryogenic thermoplastic laser labels with a glove friendly removable adhesive. Ideal for long-term storage identification of flat-surface containers in liquid nitrogen and harsh deep-freeze environments used in biomedical research and clinical laboratories. The clear cryo labels accept desktop laser printing and permanent marker. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.
Catalog No. 50-005-71867 Supplier GA International Supplier No. TRCL6
Transparent cryogenic thermoplastic laser labels with a glove friendly removable adhesive. Ideal for long-term storage identification of flat-surface containers in liquid nitrogen and harsh deep-freeze environments used in biomedical research and clinical laboratories. The clear cryo labels accept desktop laser printing and permanent marker. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.