Learn More
GA International Transparent Removable Cryogenic Labels for Laser Printers 2" x 2" / 50.8mm x 50.8mm Rectangle Transparent From -196°C to +120°C (-321°F to +248°F)
Catalog No. 5000572561
Encompass_Preferred
Transparent cryogenic thermoplastic laser labels with a glove friendly removable adhesive. Ideal for long-term storage identification of flat-surface containers in liquid nitrogen and harsh deep-freeze environments used in biomedical research and clinical laboratories. The clear cryo labels accept desktop laser printing and permanent marker. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.
Catalog No. 50-005-72561 Supplier GA International Supplier No. TRCL20
Transparent cryogenic thermoplastic laser labels with a glove friendly removable adhesive. Ideal for long-term storage identification of flat-surface containers in liquid nitrogen and harsh deep-freeze environments used in biomedical research and clinical laboratories. The clear cryo labels accept desktop laser printing and permanent marker. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.