Learn More
Research Products International Corp Transparent Vacuum Desiccator, 5 7/8 x 8 1/8 Inches
SDP
Supplier:  Research Products International Corp 420012
Encompass
Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dry storage and protection for sensitive products.
Tapered polypropylene stopcock with PTFE plug accepts tubing with ¼ to ⅜” I.D. and allows a controlled entrance of air preventing unnecessary turbulence
Neoprene O-ring assures a tight seal down to 0˚C without the need for grease (ring will not be harmed if grease is used) ‘Flat Dome,&rsquo.WARNING  This product can expose you to chemicals including Choloprene and Bisphenol-A , which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-212-717
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov