Learn More
Electron Microscopy Sciences Microwave Vented Coplin Jars
SDP
Catalog No. 5030576
Encompass
Microwave Coplin Jars with a vented lid are ideal for use in light or electron microscopy microwave laboratory procedures..
Catalog No. 50-305-76 Supplier Electron Microscopy Sciences Supplier No. 97087
Microwave Coplin Jars with a vented lid are ideal for use in light or electron microscopy microwave laboratory procedures..