Learn More
Fisherbrand™ Transport Container
Catalog No. 14127655 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear, Red
Clear, Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-655 Clear, Yellow
14-127-654 Clear, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-127-655
Only null left
Add to Cart
Add to Cart
Designed for safe, secure transport of biological/clinical samples
  • The Fisherbrand Transport Container transparent design is OSHA 29 CFR 1910.1031 and UN3373 compliant
  • Polycarbonate construction and available in three colors for user/ owner identification
  • Accommodates 13 mm, 16 mm and 20 mm tubes with our Half and Full Size OneRack™
  • Use the separators provided to create your own compartment sizes
  • Lid Opens a Full 180°C to expedite loading and emptying
Specifications
Product Type Transport Case
Color Clear, Yellow
Material Polycarbonate
Dimensions (L x W x H) 15 x 7.7 x 6.3 in.