Learn More
Alkali Scientific ASI™ Transport/Culture Tubes, 14mL, PP
SDP
Catalog No. 503029152
Encompass_Preferred
These 16 x 100 mm screw cap test tubes are available in Culture Tubes, transparent polypropylene with a “skirted” self-standing bottom or in crystal clear polystyrene with a traditional round bottom. Each tube has easy to read graduations and a “frosted” white marking area (12x50mm). Packed 100 tubes (with attached caps) per sterile bag, 10 bags per unit. Leak-proof screw caps White printed graduations Sterile bags of 100 tubes White marking spot Self-standing / round bottom
Catalog No. 50-302-9152 Supplier Alkali Scientific Supplier No. CT3050
These 16 x 100 mm screw cap test tubes are available in Culture Tubes, transparent polypropylene with a “skirted” self-standing bottom or in crystal clear polystyrene with a traditional round bottom. Each tube has easy to read graduations and a “frosted” white marking area (12x50mm). Packed 100 tubes (with attached caps) per sterile bag, 10 bags per unit. Leak-proof screw caps White printed graduations Sterile bags of 100 tubes White marking spot Self-standing / round bottom