Learn More
Med Vet International Specimen Collection and Transport System BD™ Sterile, 220531, 50/Pack
Catalog No. 503044367
Encompass_Preferred
Sustains viability for viruses, chlamydiae, mycoplasmas and ureaplasmas in a single formulation Utilizes a phenol red pH indicator to ensure medium integrity at the time of specimen collection Flat-bottomed conical tube stands on the benchtop which reduces the risk of biohazardous spills Capture-cap on 3 mL vial eliminates the need for forceps to remove the swab and reduces the risk of contamination Scored swabs eliminate the need for scissors
Catalog No. 50-304-4367 Supplier Med Vet International Supplier No. 785246PK
Sustains viability for viruses, chlamydiae, mycoplasmas and ureaplasmas in a single formulation Utilizes a phenol red pH indicator to ensure medium integrity at the time of specimen collection Flat-bottomed conical tube stands on the benchtop which reduces the risk of biohazardous spills Capture-cap on 3 mL vial eliminates the need for forceps to remove the swab and reduces the risk of contamination Scored swabs eliminate the need for scissors