Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Transport Medium

Catalog No. MT25500CM Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
MT25500CM 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25500CM Supplier Corning™ Supplier No. 25500CM
Add to Cart
Add to Cart

Corning™ Transport Medium is an isotonic, neutral pH solution that contains protective proteins, antibiotics to prevent microbial growth and contamination

  • This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
  • For research and further manufacturing purposes only
  • Not for direct use in humans or animals.
  • Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
  • Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines
For Use With (Application) Research, Manufacturing Applications
Quantity 6/Cs.
Storage Requirements 2°C to 8°C
Volume (Metric) 1 L
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.