Learn More
Vikan™ Transport Storage Tub with Drain Plug
Catalog No. 17227670 Shop All Remco Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-227-670 White
1 options
Catalog No. 17-227-670 Supplier Vikan™ Supplier No. 69155
Add to Cart
Add to Cart
Transport Storage Tub with Drain Plug is commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications.
  • Offer a combination of durability and versatility
  • Manufactured from high-density polyethylene, they can be used for transporting, freezing, and storing
  • Tubs are available as standalone items or with low steel undercarriages
  • This tub has been altered to include a drain plug, which is not currently FDA approved for food contact
  • The addition of a plug makes it easy to drain and clean tubs, and is ideal for seafood that needs to be kept in ice
Specifications
Product Type Tub
Color White
Material Polyethylene
Certifications/Compliance FDA Compliant
Quantity 1
Dimensions (L x W x H) 46.8 x 26.5 x 16.9 in.