Learn More
Chemglass Life Sciences Transport/Mailing Tubes, 10mL, 16 x 80mm, w/Attached Screw Cap, Sterile, 10 bags of 100 tubes
SDP
Catalog No. 501979493
Encompass_Preferred
Transport/Mailing Tubes, 10mL, 16 x 80mm, w/Attached Screw Cap, Sterile, 10 bags of 100 tubes. Used to store, transport or freeze samples, these tubes come in both 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes.
Catalog No. 50-197-9493 Supplier Chemglass Life Sciences Supplier No. CLS4740010
Transport/Mailing Tubes, 10mL, 16 x 80mm, w/Attached Screw Cap, Sterile, 10 bags of 100 tubes. Used to store, transport or freeze samples, these tubes come in both 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes.