Learn More
Chemglass Life Sciences Transport/Mailing Tubes, 5mL, 16 x 60mm, w/Attached Screw Cap, Sterile, 10 bags of 100 tubes
SDP
Catalog No. 501979492
Encompass_Preferred
Transport/Mailing Tubes, 5mL, 16 x 60mm, w/Attached Screw Cap, Sterile, 10 bags of 100 tubes. Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes.
Catalog No. 50-197-9492 Supplier Chemglass Life Sciences Supplier No. CLS4740005
Transport/Mailing Tubes, 5mL, 16 x 60mm, w/Attached Screw Cap, Sterile, 10 bags of 100 tubes. Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes.