Learn More
Electron Microscopy Sciences Transport Tube Sample White caps, 5 ml, Sterile
SDP
Catalog No. 501899103
Encompass_Preferred
Transport Vials - Ideal for sample collection and storage. Made from virgin polypropylene, they are available in three sizes, and come either sterile or non-sterile. Graduations provide convenient and accurate references. Supplied with blue cap that has a molded-in sealing ring for a leak-proof fit.
Volume: 5 ml
Sterile: Sterile
Catalog No. 50-189-9103 Supplier Electron Microscopy Sciences Supplier No. 6303505
Transport Vials - Ideal for sample collection and storage. Made from virgin polypropylene, they are available in three sizes, and come either sterile or non-sterile. Graduations provide convenient and accurate references. Supplied with blue cap that has a molded-in sealing ring for a leak-proof fit.
Volume: 5 ml
Sterile: Sterile