Learn More
Electron Microscopy Sciences PS Conical Bottom with Skirt Tube No cap, 12 ml
SDP
Catalog No. 5028421
Encompass_Preferred
Transport Tubes are ideal for rapid sample collection when a plug cap is needed. ubes work well in Olympus automated testing equipment. Hollow top or hollow bottom plug caps are sold separately. Made from virgin polystyrene.
Catalog No. 50-284-21 Supplier Electron Microscopy Sciences Supplier No. 63036PS
Transport Tubes are ideal for rapid sample collection when a plug cap is needed. ubes work well in Olympus automated testing equipment. Hollow top or hollow bottom plug caps are sold separately. Made from virgin polystyrene.