Learn More
Electron Microscopy Sciences Transport Tube Sample White caps, 10 ml
SDP
Catalog No. 5028430
Encompass_Preferred
Transport Vials - Ideal for sample collection and storage. Made from virgin polypropylene, they are available in three sizes, and come either sterile or non-sterile. Graduations provide convenient and accurate references. Supplied with blue cap that has a molded-in sealing ring for a leak-proof fit.
Volume: 10 ml
Catalog No. 50-284-30 Supplier Electron Microscopy Sciences Supplier No. 6303410
Transport Vials - Ideal for sample collection and storage. Made from virgin polypropylene, they are available in three sizes, and come either sterile or non-sterile. Graduations provide convenient and accurate references. Supplied with blue cap that has a molded-in sealing ring for a leak-proof fit.
Volume: 10 ml