Learn More
JDM LAB SOLUTIONS LLC Transport Tube, Screw-cap, 10mL
Catalog No. NC4039637
Encompass
Transport Tube, non-sterile, 16 x 92mm, 10ml, with separate O-ring screw-cap, printed graduations, self-standing, 10 bags of 100 tubes and caps, 1000 per case
Catalog No. NC4039637 Supplier JDM LAB SOLUTIONS LLC Supplier No. JDMC1814NS
Transport Tube, non-sterile, 16 x 92mm, 10ml, with separate O-ring screw-cap, printed graduations, self-standing, 10 bags of 100 tubes and caps, 1000 per case