Learn More
MTC Bio Transport Tube, non-sterile, 16 x 56mm, 5ml, with separate O-Ring screw-cap, printed graduations, self-standing, 10 bags of 100 tubes and caps, 1000 per case
Catalog No. NC3737099
Encompass_Preferred
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof o-ring sealing screw-caps. They are made of high-grade polypropylene, and include molded graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes• Leak-proof, 1.25 turn screw-cap• Meet 95kPa transportation guidelines• Vivid, printed graduations• Sterile, with attached screw caps• Non-Sterile, with separately packaged screw caps• RNase, DNase, Pyrogen free
Catalog No. NC3737099 Supplier MTC Bio Supplier No. C1813NS
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof o-ring sealing screw-caps. They are made of high-grade polypropylene, and include molded graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes• Leak-proof, 1.25 turn screw-cap• Meet 95kPa transportation guidelines• Vivid, printed graduations• Sterile, with attached screw caps• Non-Sterile, with separately packaged screw caps• RNase, DNase, Pyrogen free