Learn More
MTC Bio Transport Tube, sterile, 16 x 60mm, 5ml, with screw-cap, printed graduations, self-standing, 100 tubes per bag, with caps attached, 1000/cs
Supplier:  MTC Bio C1813
Encompass_Preferred
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes. Leak-proof, 1.25 turn screw-cap. Meet 95kPa transportation guidelines. Vivid, printed graduations. Screw-caps attached. Sterile, RNase, DNase, Pyrogen free
Catalog No. 50-226-8453