Learn More
Alkali Scientific Transport Tubes with Temperature Indicator, 500/Cs, 5mL Tubes
SDP
Catalog No. 5000663189
Encompass_Preferred
Our self standing transport tubes — often referred to as mailing tubes or transport vials—are made of virgin polypropylene and are supplied with 1¼ turn screw caps attached. These tubes are certified to be leak proof and 95kPa compliant and are now available with FreezeTech™ Temperature Indicator. With FreezeTech™, each tube features a temperature indicator patch that starts turning blue at 4°C and becomes dark blue when the tube (and its contents) reach 0°C. The indicator patch returns to its original transparent grey color as the sample temperature approaches 4°C and above. This serves as a warning that critical samples may be reaching undesirable temperatures, making it the ideal vessel for transport of temperature sensitive materials. • FreezeTech™ Temperature Indicator • Meets 95kPa Transport Standards • White printed graduations & marking area • Sterile, with leak proof caps attached • RNase, DNase, and pyrogen free
Catalog No. 50-006-63189 Supplier Alkali Scientific Supplier No. C2805
Our self standing transport tubes — often referred to as mailing tubes or transport vials—are made of virgin polypropylene and are supplied with 1¼ turn screw caps attached. These tubes are certified to be leak proof and 95kPa compliant and are now available with FreezeTech™ Temperature Indicator. With FreezeTech™, each tube features a temperature indicator patch that starts turning blue at 4°C and becomes dark blue when the tube (and its contents) reach 0°C. The indicator patch returns to its original transparent grey color as the sample temperature approaches 4°C and above. This serves as a warning that critical samples may be reaching undesirable temperatures, making it the ideal vessel for transport of temperature sensitive materials. • FreezeTech™ Temperature Indicator • Meets 95kPa Transport Standards • White printed graduations & marking area • Sterile, with leak proof caps attached • RNase, DNase, and pyrogen free