Learn More
MTC Bio Transport Tube, 5mL, separate screw-cap with O-ring, molded graduations, Sterile, 10 bags of 100 tubes and caps, 1000 per case
Catalog No. NC3737095
Encompass_Preferred
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes• Leak-proof, 1.25 turn screw-cap• Meet 95kPa transportation guidelines• Vivid, printed graduations• Screw-caps attached• Sterile, RNase, DNase, Pyrogen free
Catalog No. NC3737095 Supplier MTC Bio Supplier No. C1811SC
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes• Leak-proof, 1.25 turn screw-cap• Meet 95kPa transportation guidelines• Vivid, printed graduations• Screw-caps attached• Sterile, RNase, DNase, Pyrogen free