Learn More
MTC Bio Transport Tube, sterile, 16 x 60mm, 5ml, with screw-cap, printed graduations, self-standing, 100 tubes per bag, with caps attached, 1000/cs
Catalog No. NC1859344
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes
Catalog No. NC1859344 Supplier MTC Bio Supplier No. C1811
This item has a minimum qty of 2 per supplier requirements.
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes