Learn More
JDM LAB SOLUTIONS LLC Transport Vial, Screwcap, 5mL, TempPatch
Catalog No. NC4039638
Encompass
Transport Tube, with FreezeTech™ Temperature Indicator (turns blue at 0 degrees C), 5mL, attached screw-cap, printed graduations, sterile, 5 bags of 100 tubes and caps, 500/cs
Catalog No. NC4039638 Supplier JDM LAB SOLUTIONS LLC Supplier No. JDMC1825
Transport Tube, with FreezeTech™ Temperature Indicator (turns blue at 0 degrees C), 5mL, attached screw-cap, printed graduations, sterile, 5 bags of 100 tubes and caps, 500/cs