Learn More
Electron Microscopy Sciences TRANSPORT VIALS 10 ML 16X80MM
SDP
Catalog No. 502413040
Encompass_Preferred
Transport tubes (aka "mailing tubes") are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.These tubes are made of polypropylene and are supplied with the screw-caps attached, sterile, in bags of 100 tubes. Transport Tubes are self-standing and have a white printed marking spot and graduations.
Catalog No. 50-241-3040 Supplier Electron Microscopy Sciences Supplier No. 6303802
Transport tubes (aka "mailing tubes") are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.These tubes are made of polypropylene and are supplied with the screw-caps attached, sterile, in bags of 100 tubes. Transport Tubes are self-standing and have a white printed marking spot and graduations.