Description
- Highest quality plastic injection moulding
- Every kit is shrink-wrapped reducing the risk of damage during transit, offering extra protection whilst in storage
- A cost-effective solution for the lab
- Our transportation vials are designed with an ergonomically textured cap to ensure ease of use, providing a secure and comfortable grip for effortless opening and closing
- Sturdy integrated spoon to collect patient samples
- Prefilled with Apafix, a formalin-free, non-toxic, environmentally friendly fixative that carries no warning symbols
Specifications
Specifications
| Product Type | Specimen Collection Kit |
| Format | Tube |
| Media Type | UTM-RT |
| Sterility | Non-sterile |
| Color | Green |
| Material | Polypropylene |