Learn More
Fisherbrand™ Transportation Clean Vial
Catalog No. 21051030 Shop All Fisherbrand Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
21-051-030 Non-sterile
1 options
Catalog No. 21-051-030
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Fisherbrand Transportation Vials are high-quality, cost-effective vials utilised for the collection of patient samples.
  • Highest quality plastic injection moulding
  • Every kit is shrink-wrapped reducing the risk of damage during transit, offering extra protection whilst in storage
  • A cost-effective solution for the lab
  • Our transportation vials are designed with an ergonomically textured cap to ensure ease of use, providing a secure and comfortable grip for effortless opening and closing
  • Sturdy integrated spoon to collect patient samples
Specifications
Product Type Specimen Collection Kit
Format Tube
Media Type UTM-RT
Sterility Non-sterile
Color White
Material Polypropylene