Learn More
Kyfora Bio Transporter 5 Transfection Reagent 50ML
Catalog No. 502559817
Encompass_Preferred
Kyfora Bio's Transporter 5 is a high-quality, ready-to-use transfection reagent derived from our popular PEI MAX (Polyethyleneimine MAX). This versatile reagent is suitable for both stable and transient transfections across a variety of cell lines, including HEK293 and CHO cells. Transporter 5 consistently delivers high yields of viral vectors (AAV and LV), recombinant proteins, and monoclonal antibodies, all while maintaining optimal cell viability. Its scalability and compatibility with a range of cell culture media make it an excellent choice among available transfection reagents on the market.
Catalog No. 50-255-9817 Supplier Kyfora Bio Supplier No. 2600850
Kyfora Bio's Transporter 5 is a high-quality, ready-to-use transfection reagent derived from our popular PEI MAX (Polyethyleneimine MAX). This versatile reagent is suitable for both stable and transient transfections across a variety of cell lines, including HEK293 and CHO cells. Transporter 5 consistently delivers high yields of viral vectors (AAV and LV), recombinant proteins, and monoclonal antibodies, all while maintaining optimal cell viability. Its scalability and compatibility with a range of cell culture media make it an excellent choice among available transfection reagents on the market.