Learn More
TRIBIOSCIENCE INC TransReagent (Transfection Reagent) Maxi, 1 ML
SDP
Catalog No. NC3902635
Encompass
TransReagent Maxi is a polymer-based reagent without animal derivers. It has high-efficiency, low-toxicity, strong-stability, and is suitable for many cell types including suspension cells, such as HEK-293, HEK-293T, CHO-K1, expi293, expiCHO, COS-1, COS-7, NIH/3T3, Sf9, HepG2, and other cells. It can also be applied to large-scale recombinant protein expression and virus production.
Catalog No. NC3902635 Supplier TRIBIOSCIENCE INC Supplier No. TBS804201
TransReagent Maxi is a polymer-based reagent without animal derivers. It has high-efficiency, low-toxicity, strong-stability, and is suitable for many cell types including suspension cells, such as HEK-293, HEK-293T, CHO-K1, expi293, expiCHO, COS-1, COS-7, NIH/3T3, Sf9, HepG2, and other cells. It can also be applied to large-scale recombinant protein expression and virus production.