Learn More
Athens Research And Technology Inc TRANSTHYRETIN HUMAN 1MG
Supplier:  Athens Research And Technology Inc 1616161801TRY1MG
Encompass_Preferred
Transthyretin, Human Plasma (Prealbumin): 1 MG; Lyophilized from 50 mM Na phosphate, pH 7.5, with 150 mM NaCl.; MW 55,000; EC= 1.35; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from human plasma.
Catalog No. 50-489-933