Learn More
MyBioSource Human TTR ELISA Kit
SDP
Supplier:  MyBioSource MBS201893896STRIPWELLS
Encompass_Preferred
The MyBioSource Human TTR ELISA Kit quantifies human transthyretin in various biological fluids using a 96-well plate coated with a TTR-specific antibody. After binding and washing steps, a biotinylated antibody and HRP-conjugated streptavidin are added. TMB substrate reveals TTR levels by color change, measured at 450 nm.
Catalog No. NC2306750