Learn More
Medchemexpress LLC Transthyretin/TTR H 2ug
SDP
Catalog No. 5000196006
Encompass_Preferred
Recombinant human transthyretin (TTR) expressed in HEK293 cells with C-terminal human Fc and C-Myc tags. Supplied as a purified protein suitable for biochemical assays, interaction studies, and research into amyloidosis and Alzheimer's disease. Packaged at 2 μg for small-scale experiments.
  • Human TTR expressed in mammalian cells for native folding and post-translational modifications.
  • C-terminal Fc and Myc tags to facilitate detection and purification.
  • Suitable for biochemical assays and protein-protein interaction studies.
  • Useful for amyloidosis and Alzheimer's disease research models.
  • Supplied in a small pack size for pilot or optimization experiments.
Catalog No. 50-001-96006 Supplier Medchemexpress LLC Supplier No. HYP720262UG
Recombinant human transthyretin (TTR) expressed in HEK293 cells with C-terminal human Fc and C-Myc tags. Supplied as a purified protein suitable for biochemical assays, interaction studies, and research into amyloidosis and Alzheimer's disease. Packaged at 2 μg for small-scale experiments.
  • Human TTR expressed in mammalian cells for native folding and post-translational modifications.
  • C-terminal Fc and Myc tags to facilitate detection and purification.
  • Suitable for biochemical assays and protein-protein interaction studies.
  • Useful for amyloidosis and Alzheimer's disease research models.
  • Supplied in a small pack size for pilot or optimization experiments.