Learn More
ACROBiosystems Recombinant Protein;Human Transthyretin / Prealbumin Protein;HEK293;100UG;TTR,Transthyretin,TTR,Prealbumin,TBPA,ATTR,PALB,CTS,CTS1,HsT2651
Catalog No. 501658435
Encompass_Preferred
Recombinant Protein;Human Transthyretin / Prealbumin Protein;100UG;Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession # NP_000362).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.6 kDa. The protein migrates as 19 kDa and 37kDa under reducing (R) condition (SDS-PAGE) due to the monomer and dimer respectively.
Catalog No. 50-165-8435 Supplier ACROBiosystems Supplier No. TTRH5223100UG
Recombinant Protein;Human Transthyretin / Prealbumin Protein;100UG;Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession # NP_000362).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.6 kDa. The protein migrates as 19 kDa and 37kDa under reducing (R) condition (SDS-PAGE) due to the monomer and dimer respectively.