Learn More
Enzo Life Sciences Tranylcypromine (1g). CAS: 13492-01-8
SDP
Supplier:  Enzo Life Sciences BMLEI2170001
Encompass_Preferred
Inhibits prostacyclin synthase. Monoamine oxidase inhibitor. Effective small molecule inhibitor of histone demethylation. Inhibits the demethylase BHC110/LSD1 (IC50<2μM). When combined with a GSK-3 inhibitor, it causes reprogramming of human primary keratinocyte transduced with Oct4 and Klf4. Alternative name: Trans(+/-)-2-phenylcyclopropanamine, SKF trans-355, Tranylcypromine hemisulfate , 2-PCPA. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0614