Learn More
Medchemexpress LLC HY-B1496 100mg Medchemexpress, Tranylcypromine (hemisulfate) CAS:13492-01-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1496100MG
Encompass_Preferred
Medchemexpress, HY-B1496 100mg Tranylcypromine (hemisulfate) CAS:13492-01-8 Tranylcypromine hemisulfate is an irreversible, nonselective MAO inhibitor used in the treatment of depression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2955