Learn More
Medchemexpress LLC Tranylcypromine hemisulfate | 13492-01-8 | 99.9% | 1 ML
SDP
Catalog No. 5000411685
Encompass_Preferred
Tranylcypromine hemisulfate is an irreversible, nonselective monoamine oxidase (MAO) inhibitor used in the treatment of depression. It also acts as a lysine-specific demethylase 1 (LSD1) inhibitor, capable of suppressing lesion growth and ameliorating generalized hyperalgesia in mouse models of induced endometriosis. This compound exhibits antidepressant effects.
  • Irreversible, nonselective monoamine oxidase (MAO) inhibitor.
  • Lysine-specific demethylase 1 (LSD1) inhibitor.
  • Suppresses lesion growth and ameliorates generalized hyperalgesia.
  • Neuroprotective against human Aβ(1-42) oligomer-induced toxicity.
  • Protects RGCs from glutamate neurotoxicity-induced apoptosis and oxidative stress.
  • Promotes mitogen-activated protein kinase 12 (p38 MAPKγ) expression.
  • Enhances RGC survival by attenuating NMDA neurotoxicity.
  • Increases cell proliferation in brain regions.
Catalog No. 50-004-11685 Supplier Medchemexpress LLC Supplier No. HYB149610MM/1ML
Tranylcypromine hemisulfate is an irreversible, nonselective monoamine oxidase (MAO) inhibitor used in the treatment of depression. It also acts as a lysine-specific demethylase 1 (LSD1) inhibitor, capable of suppressing lesion growth and ameliorating generalized hyperalgesia in mouse models of induced endometriosis. This compound exhibits antidepressant effects.
  • Irreversible, nonselective monoamine oxidase (MAO) inhibitor.
  • Lysine-specific demethylase 1 (LSD1) inhibitor.
  • Suppresses lesion growth and ameliorates generalized hyperalgesia.
  • Neuroprotective against human Aβ(1-42) oligomer-induced toxicity.
  • Protects RGCs from glutamate neurotoxicity-induced apoptosis and oxidative stress.
  • Promotes mitogen-activated protein kinase 12 (p38 MAPKγ) expression.
  • Enhances RGC survival by attenuating NMDA neurotoxicity.
  • Increases cell proliferation in brain regions.