Learn More
Cayman Chemical Tranylcypromine hydrochlorid
SDP
Catalog No. 502887685
Encompass_Preferred
An irreversible, mechanism-based inhibitor of LSD1 (IC50 = 20.7 µM); irreversibly inhibits MAO-A and MAO-B (IC50s = 2.3 and 0.95 µM, respectively); PCPA inhibits nucleosomal demethylation of H3K4 (IC50 = <2 µM); induces EpiSCs to convert to more undifferentiated state but requires inhibitors of ALK5, MEK, FGFR, and GSK3 for full conversion to pluripotent mESCs; decreases proliferation of neural progenitors in the hippocampal dentate gyrus in adult mice at 10 mg/kg for ten days; decreases the time juvenile rats spend immobile in the forced swim test at 0.4-10 mg/kg
Catalog No. 50-288-7685 Supplier Cayman Chemical Supplier No. 10010494500 MG
An irreversible, mechanism-based inhibitor of LSD1 (IC50 = 20.7 µM); irreversibly inhibits MAO-A and MAO-B (IC50s = 2.3 and 0.95 µM, respectively); PCPA inhibits nucleosomal demethylation of H3K4 (IC50 = <2 µM); induces EpiSCs to convert to more undifferentiated state but requires inhibitors of ALK5, MEK, FGFR, and GSK3 for full conversion to pluripotent mESCs; decreases proliferation of neural progenitors in the hippocampal dentate gyrus in adult mice at 10 mg/kg for ten days; decreases the time juvenile rats spend immobile in the forced swim test at 0.4-10 mg/kg