Learn More
Medchemexpress LLC Tranylcypromine hydrochloride | 1986-47-6 | 99.91% | 169.66 | 100 MG
SDP
Catalog No. 5000386252
Encompass_Preferred
Tranylcypromine hydrochloride is an irreversible inhibitor of lysine-specific demethylase 1 (LSD1/BHC110) and monoamine oxidase (MAO). It effectively inhibits LSD1, MAO A, and MAO B with specified IC50s. This compound is utilized for research applications, particularly those related to depression.
  • Irreversible inhibitor of lysine-specific demethylase 1 (LSD1/BHC110).
  • Irreversible inhibitor of monoamine oxidase (MAO).
  • Inhibits LSD1 with an IC50 of 20.7 μM.
  • Inhibits MAO A with an IC50 of 2.3 μM.
  • Inhibits MAO B with an IC50 of 0.95 μM.
  • Suitable for research related to depression.
  • Supplied as a white to yellow solid.
  • Requires sealed storage at 4°C, away from moisture.
  • Shows specific derepression of OCT4 transcription in vitro.
  • Decreases LPS-mediated microglial activation in vivo.
  • Down-regulates Aβ-mediated microglial activation in vivo.
Catalog No. 50-003-86252 Supplier Medchemexpress LLC Supplier No. HY17447AG100MG
Tranylcypromine hydrochloride is an irreversible inhibitor of lysine-specific demethylase 1 (LSD1/BHC110) and monoamine oxidase (MAO). It effectively inhibits LSD1, MAO A, and MAO B with specified IC50s. This compound is utilized for research applications, particularly those related to depression.
  • Irreversible inhibitor of lysine-specific demethylase 1 (LSD1/BHC110).
  • Irreversible inhibitor of monoamine oxidase (MAO).
  • Inhibits LSD1 with an IC50 of 20.7 μM.
  • Inhibits MAO A with an IC50 of 2.3 μM.
  • Inhibits MAO B with an IC50 of 0.95 μM.
  • Suitable for research related to depression.
  • Supplied as a white to yellow solid.
  • Requires sealed storage at 4°C, away from moisture.
  • Shows specific derepression of OCT4 transcription in vitro.
  • Decreases LPS-mediated microglial activation in vivo.
  • Down-regulates Aβ-mediated microglial activation in vivo.