Learn More
CPC Scientific H-Ser-Phe-Leu-Leu-Arg-NH2 (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific PARP017A
Encompass
Thrombin receptor agonist peptide.SEQUENCE: H-Ser-Phe-Leu-Leu-Arg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: SFLLR-NH2MOLECULAR FORMULA: C30H51N9O6MOLECULAR WEIGHT: 633.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [141923-41-3]SYNONYMS: Proteinase Activated Receptor 1 (1-5) amide (human), SFLLRamide, PAR-1 (1-5) amide (human), Coagulation Factor II Receptor (1-5) amide (human), Thrombin Receptor (1-5) amide (human), Thrombin Receptor Activator Peptide 5 amideRESEARCH AREA: HematologyREFERENCES:R.M.Scarborough et al., J. Biol. Chem., 267, 13146 (1992)SKU(s): PARP-017A
Catalog No. 50-211-0124