Learn More
CPC Scientific H-Phe-Leu-Leu-Arg-Asn-OH (trifluoroacetate salt) 5MG
SDP
Catalog No. 502110129
Encompass
FLLRN inhibits platelet aggregation induced by SFLLRN or thrombin. This peptide is a PAR 1 antagonist peptide.SEQUENCE: H-Phe-Leu-Leu-Arg-Asn-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: FLLRNMOLECULAR FORMULA: C31H51N9O7MOLECULAR WEIGHT: 661.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [141136-84-7]SYNONYMS: TRAP6 (2-6)RESEARCH AREA: HematologyREFERENCES:Al-Ani, B. et al. Br. J. Pharmacol. 128, 1105 (1999)Matsoukas, J. et al. J. Protein Chem. 16, 113 (1997)SKU(s): PARP-020A, PARP-020B
Catalog No. 50-211-0129 Supplier CPC Scientific Supplier No. PARP020A
FLLRN inhibits platelet aggregation induced by SFLLRN or thrombin. This peptide is a PAR 1 antagonist peptide.SEQUENCE: H-Phe-Leu-Leu-Arg-Asn-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: FLLRNMOLECULAR FORMULA: C31H51N9O7MOLECULAR WEIGHT: 661.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [141136-84-7]SYNONYMS: TRAP6 (2-6)RESEARCH AREA: HematologyREFERENCES:Al-Ani, B. et al. Br. J. Pharmacol. 128, 1105 (1999)Matsoukas, J. et al. J. Protein Chem. 16, 113 (1997)SKU(s): PARP-020A, PARP-020B