Learn More
Bachem TRAP-6 amide Trifluoroacetate
SDP
Catalog No. 50259669
Encompass_Preferred
TRAP-6 amide Trifluoroacetate, H-Ser-Phe-Leu-Leu-Arg-Asn-NH2, 141923-40-2, C34H57N11O8, Mr 747.90, 25mg. PAR-1 (1-6) amide (human), Thrombin Receptor Activator Peptide 6 amide, Proteinase Activated Receptor 1 (1-6) amide (human), Thrombin Receptor (1-6) amide (human), Coagulation Factor II Receptor (1-6) amide (human), SFLLRNamide, SFLLRN-NH2 Selective Protease-Activated Receptor 1 (PAR1) Agonist. SFLLRN-amide, thrombin receptor agonist peptide.
Catalog No. 50-259-669 Supplier Bachem Supplier No. 4031274.0025
TRAP-6 amide Trifluoroacetate, H-Ser-Phe-Leu-Leu-Arg-Asn-NH2, 141923-40-2, C34H57N11O8, Mr 747.90, 25mg. PAR-1 (1-6) amide (human), Thrombin Receptor Activator Peptide 6 amide, Proteinase Activated Receptor 1 (1-6) amide (human), Thrombin Receptor (1-6) amide (human), Coagulation Factor II Receptor (1-6) amide (human), SFLLRNamide, SFLLRN-NH2 Selective Protease-Activated Receptor 1 (PAR1) Agonist. SFLLRN-amide, thrombin receptor agonist peptide.